Mini Dissecting Scissors, Straight, 9.5 cm | Avantor
Mini Dissecting Scissors, Straight, 9.5 cm
Mini Dissecting Scissors, Straight, 9.5 cm
Certificates
Mini Dissecting Scissors, Straight, 9.5 cm
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Our mini dissecting scissors with Large Rings are similar to regular dissecting scissors, but they are shorter and have more delicate tips, making them ideal for fine dissection work in tight spaces. They are commonly used in microsurgery, like ophthalmic surgery. These dissecting scissors have large rings for people with larger fingers.
  • These scissors are 9.5 cm (3.75") long
  • Straight blades with rounded tips and fine blades
These dissecting scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. (Standard and medium blades are also available on our mini dissecting scissors).
Dissecting scissors have thinner (and often longer) blades than operating scissors. They are designed for cutting and probing in tight spaces. Their tips are pointed, where operating scissors are generally blunt. These are available in surgical grade stainless steel.
These dissecting scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. To guarantee a long life for your surgical instruments, be sure to clean them thoroughly after each use.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Curvature
    Handle
    Availability
    Price
  • Dissecting miniature scissors, fine, round tips
    Surgical grade stainless steel
    Straight
    Square ring handles
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $99.78
    $99.78
    Each
       $99.78
    Supplier Number: 503242
  • Dissecting miniature scissors, regular size tips
    Surgical grade stainless steel
    Straight
    Ring handles
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $99.78
    $99.78
    Each
       $99.78
    Supplier Number: 503244
  • Mini dissecting scissors, fine, sharp round tips
    Surgical grade stainless steel
    Straight
    Large ring handles for people with larger fingers
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $133.83
    $133.83
    Each
       $133.83
    Supplier Number: 503245
  • Mini dissecting scissors, fine, sharp tips
    Surgical grade stainless steel
    Straight
    Ring handles
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $99.78
    $99.78
    Each
       $99.78
    Supplier Number: 503241
  • Mini dissecting scissors, medium width blades, fine, sharp tips
    Surgical grade stainless steel
    Straight
    Ring handles
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $99.78
    $99.78
    Each
       $99.78
    Supplier Number: 503243
Certains produits peuvent apparaître comme restreints
Pour garantir un passage en caisse fluide et rapide,veuillez-vous connecter à votre compte.Certains articles peuvent apparaître comme restreints simplement parce que vous n'êtes pas connecté.
Si vous n'avez pas de compte, vous pouvez vous inscrire en utilisant notre formulaire d'inscription en ligne (https://www.avantorsciences.com/ca/en/login/register)
Si des restrictions apparaissent toujours après votre connexion, certains produits — comme les produits chimiques ou les dispositifs médicaux — nécessitent des étapes supplémentaires de vérification de compte pour pouvoir passer une commande. Certains articles peuvent également nécessiter une licence spécifique ou une documentation supplémentaire ; la documentation requise sera demandée pour ces articles avant l’expédition.
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.