Reference Cell Electrodes | Avantor
Reference Cell Electrodes
Reference Cell Electrodes
Certificates
Reference Cell Electrodes
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
New, improved sintered pellets with lower resistance and high strength. Stable and well balanced in the presence of current, these small and inexpensive half-cells are easy to work with as bath electrodes.
  • Reference cell electrodes for laboratory research
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Reference cell with 4' lead
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $180.12
    $180.12
    Each
       $180.12
    Supplier Number: RC1
  • Reference cell with 4' lead, 2.0 mm pin
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $180.12
    $180.12
    Each
       $180.12
    Supplier Number: RC1T
  • Reference cell with 2.0 mm pin
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $132.72
    $132.72
    Each
       $132.72
    Supplier Number: RC2
  • Reference cell with female connector
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $164.32
    $164.32
    Each
       $164.32
    Supplier Number: RC2F
  • Reference cell with epoxy body, 50 mm long, Ø 4.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $205.40
    $205.40
    Each
       $205.40
    Supplier Number: RC3
  • Reference cell with glass body, 50 mm long, Ø 1.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $287.56
    $287.56
    Each
       $287.56
    Supplier Number: RC6