Dual Position Ventilation Caps
77588-232
New Product
- Description:Dual position closure cap
- Material:LDPE
- Cat. no.:77588-232
- For Tube O.D.:16 mm
- Cap Color:Neutral
High-quality, safe, and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
- Two position fit allows both gas exchange and sealing
- Ribbed surface for maximum grip
Dual position ventilation caps offer excellent sealing properties, and are available in LDPE material.