Screw Caps for Screw Cap Micro Tubes
New Product
High-quality, safe, and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
- Polypropylene material
- O-ring increases cap security (IATA tested)
- Different cap colors for better coding
- Sterile double bag packaging prevents contamination
- Ribbed surface for maximum grip
- Can be used on all SARSTEDT screw cap micro tubes
Screw caps offer excellent sealing properties, and are available in a wide variety of colors to facilitate organization and coding.
Assorted color cap orders include Yellow, Pink, Orange, Red, Purple, Green, Blue, Brown, Black, and Neutral colors. Sterile and nonsterile versions available.
Frequently Bought Together