Skirted Push Caps
Fournisseur: SARSTEDT INC
New Product
Certificats
À propos de cet article
High-quality, safe, and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
- Protruding skirt prevents contamination
- Ribbed surface for maximum grip
Skirted push caps offer excellent sealing properties, and are available in LDPE or HDPE material.
Frequently Bought Together