Group Lockout Box, Brady®
40.6 x 6.4 x 19 cm (71/2 x 16 x 21/2 x 71/2")
Catalog # CA82004-262
Supplier: Avantor
Jump to:
Product Details & Documents
This lockout center features 18 hooks for keys and 2 slots for locks (keys and locks not included)
The box allows each person to apply a padlock to the key box effectively allowing them to lock out multiple sources of energy controlled by the padlock keys inside the box. Made of acrylic. Not designed to prevent forced entry.
Spécifications
- W×D×H:40.6 x 6.4 x 19 cm (71/2 x 16 x 21/2 x 71/2")