About this item
These microprocessor-controlled forced air units are equipped with a blow-out back panel to protect the user from injury.
- Microprocessor control system
- Over-temperature safety limit, manual restart
- Rear blow-out panel
- Heavy-gauge steel construction for durability
- VWR® Two-Year Limited Parts and Labor Warranty
The panel is designed to release in the event of an explosion, directing the force away from the user
Other features include electronic timer, heavy-duty adjustable door latch and hinges, and 7.6 cm (3") diameter exhaust port with adjustable damper. All ovens are manufactured using heavy gauge steel exteriors with continuous welded seams and corners for long-term durability. The powder coat enamel exterior has a high resistance to moisture, oil and chemicals.
Unfiltered power exhaust accessory (52201-344) is available for fast drying, to exhaust fumes from the lab, or to pull moisture out of high-temperature samples. Can be operated at the end of program for fast cool down, or during all or part of program. If attached, power exhaust can be activated by the controller.
Caution: Unit is not explosion-proof and will not prevent combustibles from exploding. The rear blow-out panel is only an added safety measure. Always use extreme caution when mixing heat and any source of volatile substances in any oven. Use with any volatile substance, material, or residual fumes beyond flashpoint may cause substances to ignite while placed in any oven. Check with your chemical source to ensure safe operational temperature ranges and mixtures while using in conjunction with any heat source. Always exercise extreme caution when handling combustible materials.
Specifications
- Catalog No:
- 52201-216
- 52201-214
- Temperature control:
- 0.05°C
- 0.05°C
- Temperature range:
- 40 to 240°C (104 to 464°F)
- 40 to 240°C (104 to 464°F)
- Temperature uniformity:
- ±1.5° @ 110°C, ±1.9° @ 170°C
- ±1.5° @ 110°C, ±1.9° @ 170°C
- Recovery Time:
- 5 min. @ 110°C (230°F), 8 min. @ 170°C (338°F)
- 5 min. @ 110°C (230°F), 8 min. @ 170°C (338°F)
Specifications
Related Information
White Glove Installation and Verification Service–the white glove installation includes unpacking, inspection, removal/disposal of packing materials, transporting order via elevator and merchandise set up in the room of your choice, installation of casters or leveling feet, power and electrical connections inspected, supply voltage verified, functional and performance verification documented. Our engineers will move the unit from the customer's receiving dock to the lab within the same building. This service does NOT include transportation from vendor warehouse to customer's receiving dock.