À propos de cet article
Full micro perforated trays with lids manufactured of stainless steel grade AISI-30.
- Electropolished
- Full wall perforation to protect sharp instruments
- Protects edges and tips from damage
- Smooth rounded corners
- Perforation hole size is 2.0 mm
- Hinged lids
Top and lid frame rod diameter: 3.5 mm and 4.0 mm (depending on the basket). Full wall perforation to protect sharp instrument's edges and tips from damage. Fine mesh with opening squares of 1.6 mm and wire diameter of 0.7 mm. Sheet locking clip.
Applications:
Micro surgical instrument cleaning and sterilization
Micro surgical instrument storage
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.