Microelectrode Holders
Microelectrode Holders
Microelectrode Holders
Certificates
Microelectrode Holders
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters.
  • Male connector
  • Wire half-cell
  • Male luer
  • Screw cap
WPI's microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers. A Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets. The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubber gaskets (without caps). Filling WPI microelectrode holders with electrolytes containing chloride results in stable electrode potential. Suitable electrolytes include KCl, NaCl and CaCl₂. Holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters.
Product Family Options
Product Information
  • For Use With
    Availability
    Price
  • 1.0 mm standard WPI single capillary tubing
    Log in for availability
    Log in for availability
    Each
    Each - $200.68
    $200.68
    Each
       $200.68
    Supplier Number: MEH2RW10
  • 1.2 mm standard WPI single capillary tubing
    Log in for availability
    Log in for availability
    Each
    Each - $200.68
    $200.68
    Each
       $200.68
    Supplier Number: MEH2RW12
  • 1.5 mm standard WPI single capillary tubing
    Log in for availability
    Log in for availability
    Each
    Each - $200.68
    $200.68
    Each
       $200.68
    Supplier Number: MEH2RW15