Ag/AgCl Pellet Electrodes
Ag/AgCl Pellet Electrodes
Ag/AgCl Pellet Electrodes
Sintered pellets with lower resistance and high strength. These small and inexpensive half-cells are easy to work with as bath electrodes.
  • Silver/Silver chloride half cells ideal for use as bath electrodes
  • Low resistance and high strength
  • Stable and well balanced in the presence of current
Product Family Options
  • Désignation
    Ø
    Longueur
  • Ag/AgCl Electrode, 12 mm long silver wire, 0.250 μm wire diameter
    12.0 mm
    1 mm
    Each
    Each - $113.78
    $113.78
    Each
       $113.78
     : EP12
  • Ag/AgCl Electrode, 12 mm long silver wire, 0.250 μm wire diameter
    4.0 mm
    1 mm
    Each
    Each - $113.78
    $113.78
    Each
       $113.78
     : EP4
  • Ag/AgCl Electrode, 12 mm long silver wire, 0.250 μm wire diameter
    8.0 mm
    1 mm
    Each
    Each - $113.78
    $113.78
    Each
       $113.78
     : EP8
  • Ag/AgCl Electrode, 70 mm long silver wire, 0.250 μm wire diameter
    1.0 mm
    3 mm
    Each
    Each - $113.78
    $113.78
    Each
       $113.78
     : EP1
  • Ag/AgCl Electrode, 5 mm long silver wire, 0.250 μm wire diameter
    2.0 mm
    4 mm
    Each
    Each - $113.78
    $113.78
    Each
       $113.78
     : EP2
  • Ag/AgCl Electrode, 40 mm long silver wire, 0.250 μm wire diameter
    0.5 mm
    20 mm
    Each
    Each - $113.78
    $113.78
    Each
       $113.78
     : EP05
  • Ag/AgCl Electrode, 40 mm long silver wire, 0.250 μm wire diameter
    0.8 mm
    20 mm
    Each
    Each - $113.78
    $113.78
    Each
       $113.78
     : EP08