Faraday bags blocks all wireless signals to protect phone data and prevent distractions.
Triple-layer CYBER NC (Nickel/Copper) fabric provides >85 dB signal attenuation
- Blocks 5G, Wi-Fi, bluetooth, GPS, RFID, and cellular signals
- Secure closures include Velcro or magnetic seals for complete signal isolation
- Durable, water-resistant exteriors in canvas or CORDURA® nylon
- ID pockets and universal sizing for school, forensic, or personal use
This collection of Faraday phone bags, NX3, PhonX3, JACKET, and JACKET Pro, offers a robust solution for blocking wireless signals and securing mobile devices in any setting. Each bag uses CYBER NC (Nickel/Copper) shielding fabric to achieve over 85 dB of attenuation across a wide frequency range (400 MHz to 40 GHz), effectively isolating devices from all common signals, including 5G, Wi-Fi, GPS, Bluetooth, RFID, and standard cellular networks.
In classrooms, these bags help minimize distractions and promote digital discipline during tests or sensitive discussions. They also provide a hands-on tool for lessons in physics and cybersecurity, illustrating electromagnetic shielding in action. From durable canvas to military-grade CORDURA® nylon, and from Velcro to magnetic closures, each model is tailored for distinct needs, from everyday classroom management to professional forensic use.