Spencer Stitch Scissors | Avantor
Spencer Stitch Scissors
Spencer Stitch Scissors
Certificates
Spencer Stitch Scissors
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Spencer Stitch Scissors are stitch-cutting scissors or suture scissors for removing sutures or stitches. Spencer Stitch Scissors have a relatively short, straight, and narrow blades.
  • Stich removal scissors
  • 9.0 to 13.25 cm long
  • Straight, blunt tips
  • Notched blade for a sure grip of the suture to cut
  • Surgical grade stainless steel or German steel
The tips of Spencer stitch scissors are blunt rather than sharp to minimize the risk of accidentally puncturing or damaging underlying tissues while cutting sutures. One blade is notched to provide improved grip and traction when cutting through sutures, preventing slippage and ensuring clean cuts of just the suture. The ring handles give you a firm, comfortable grip for precision control.
They come in a variety of lengths, from 9.0 to 13.25 cm. Shorter handles typically offer more control, and longer handles give you greater reach. These Spencer Stitch Scissors are manufactured from surgical grade stainless steel or premium German steel, both of which are autoclavable.
Recommended a properly cleaning of all your surgical instruments promptly after use. This improves the useable lifespan of your surgical tools.
Product Family Options
Informations sur le produit
  • Availability
    Price
  • Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $73.03
    $73.03
    Each
       $73.03
    Supplier Number: 500380
  • Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $113.78
    $113.78
    Each
       $113.78
    Supplier Number: 500380-G
  • Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $85.16
    $85.16
    Each
       $85.16
    Supplier Number: 501703
  • Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $120.06
    $120.06
    Each
       $120.06
    Supplier Number: 501703-G
  • Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $92.46
    $92.46
    Each
       $92.46
    Supplier Number: 501704
  • Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $113.78
    $113.78
    Each
       $113.78
    Supplier Number: 501704-G