Microelectrode Holders
Microelectrode Holders
Certificates
Microelectrode Holders
12 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
WPI™s microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers.
  • Male connector
  • Pellet half-cell
  • No pressure port
  • Screw cap
Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets. The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubber gaskets (without caps).
Filling WPI microelectrode holders with electrolytes containing chloride results in stable electrode potential. Suitable electrolytes include KCl, NaCl and CaCl₂. The holder style selected will depend on the experimental application, space, and instrumentation.
 : All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters.
Product Family Options
MPH1
Product Information
  • Désignation
    Availability
    Price
  • Microelectrode holder, size: 1.0 mm
    Log in for availability
    Log in for availability
    Each
    Each - $312.84
    $312.84
    Each
       $312.84
    Supplier Number: MPH110
  • Microelectrode holder, size: 1.2 mm
    Log in for availability
    Log in for availability
    Each
    Each - $312.84
    $312.84
    Each
       $312.84
    Supplier Number: MPH112
  • Microelectrode holder, size: 1.5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $312.84
    $312.84
    Each
       $312.84
    Supplier Number: MPH115
  • Microelectrode holder, size: 2.0 mm
    Log in for availability
    Log in for availability
    Each
    Each - $312.82
    $312.82
    Each
       $312.82
    Supplier Number: MPH120
MPH3
Product Information
  • Désignation
    Availability
    Price
  • Microelectrode holder, size: 1.0 mm
    Log in for availability
    Log in for availability
    Each
    Each - $131.14
    $131.14
    Each
       $131.14
    Supplier Number: MPH310
  • Microelectrode holder, size: 1.2 mm
    Log in for availability
    Log in for availability
    Each
    Each - $143.78
    $143.78
    Each
       $143.78
    Supplier Number: MPH312
  • Microelectrode holder, size: 1.5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $143.78
    $143.78
    Each
       $143.78
    Supplier Number: MPH315
  • Microelectrode holder, size: 2.0 mm
    Log in for availability
    Log in for availability
    Each
    Each - $143.78
    $143.78
    Each
       $143.78
    Supplier Number: MPH320
MPH4
Product Information
  • Désignation
    Availability
    Price
  • Microelectrode holder, size: 1.0 mm
    Log in for availability
    Log in for availability
    Each
    Each - $143.78
    $143.78
    Each
       $143.78
    Supplier Number: MPH410
  • Microelectrode holder, size: 1.2 mm
    Log in for availability
    Log in for availability
    Each
    Each - $143.78
    $143.78
    Each
       $143.78
    Supplier Number: MPH412
  • Microelectrode holder, size: 1.5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $143.78
    $143.78
    Each
       $143.78
    Supplier Number: MPH415
  • Microelectrode holder, size: 2.0 mm
    Log in for availability
    Log in for availability
    Each
    Each - $143.78
    $143.78
    Each
       $143.78
    Supplier Number: MPH420