Microelectrode Holders, MEH3SW
Microelectrode Holders, MEH3SW
Certificates
Microelectrode Holders, MEH3SW
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers. All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters.
  • Male connector
  • Wire half-cell
  • No pressure port
  • Screw cap
A Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets. The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubber gaskets (without caps). Filling WPI microelectrode holders with electrolytes containing chloride results in stable electrode potential. Suitable electrolytes include KCl, NaCl and CaCl₂.
Determine the required electrical connection on the holder: for example, to connect the holder to a 2 mm pin you should select a holder equipped with a 2 mm jack. Most WPI probes require a holder equipped with a 2 mm jack. Decide on the required alignment of the electrical connection: either in-line with the glass pipette, or at a right angle to it. Space considerations in your experimental setup and requirements imposed by other pieces of equipment typically determine which alignment is appropriate. Rubber gaskets offer easier insertion and removal of glass pipettes whereas screw-caps provide more secure mounts for micropipetters.
Choose a holder with either a silver wire or a silver/silver chloride pellet for the metal/liquid coupling. Silver/silver chloride pellets provide a more stable low-noise baseline which is important for low-noise DC recording. Pellets require the glass pipette and holder to be free of air bubbles to achieve a good connection. Silver wire holders are durable and are easier to use when the holder is equipped with a pressure port because the fluid in the pipette does not have to be filled to the top of the pipette to achieve a good electrical connection.
Choose a holder equipped with a pressure port only when you want to pressure inject liquid from the pipette. Two types of ports are available: 2.0 mm ext.Ø. and standard 'syringe-style' luer. The luer port is often recommended because it makes assembly and disassembly much easier. Quick-connect luer fittings for four common sizes of tubing (¹/₁₆, ³/₃₂, ¹/₈ and ⁵/₃₂" int.Ø.) are included with each luer-equipped holder.
Some non-WPI preamplifiers or headstages cannot be mounted on micromanipulators. In such cases, a holder equipped with a rod permits the holder to be conveniently mounted on a micromanipulator.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Glass microelectrode holder, 1.0 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $142.18
    $142.18
    Each
       $142.18
    Supplier Number: MEH3SW10
  • Glass microelectrode holder, 1.2 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $142.20
    $142.20
    Each
       $142.20
    Supplier Number: MEH3SW12
  • Glass microelectrode holder, 1.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $142.18
    $142.18
    Each
       $142.18
    Supplier Number: MEH3SW15
  • Glass microelectrode holder, 2.0 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $142.18
    $142.18
    Each
       $142.18
    Supplier Number: MEH3SW20