Noyes Micro Scissors, 14 cm
Noyes Micro Scissors, 14 cm
Noyes Micro Scissors, 14 cm
Certificates
Noyes Micro Scissors, 14 cm
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Noyes Micro Scissors are a surgical instrument designed for ophthalmic surgery and are an ideal choice for microdissection or for delicate small animal surgeries. Named after their inventor, ophthalmologist Dr. Arthur Noyes, the Noyes Micro Scissors are characterized by their small size and fine tips. They let you make precise and controlled incisions.
  • These scissors are 14 cm (5.5") long and come with straight or curved tips
  • Spring handle
  • Fine tips for precision surgical applications
These surgical scissors are typically used for delicate tasks such as cutting fine structures, trimming tissue, or creating small openings. Noyes scissors are a type of spring scissors which can be used ambidextrously. For this reason, Noyes scissors are often preferred by left-handed surgeons.
Noyes Micro Scissors are made from surgical grade stainless steel, which is suitable for autoclaving or chemical sterilization. Always clean your surgical instruments promptly after you use them. Proper cleaning lengthens the usable lifespan of surgical instruments.
Product Family Options
Product Information
  • Curvature
    Tip Size
    Availability
    Price
  • Straight
    20 mm
    Log in for availability
    Log in for availability
    Each
    Each - $228.72
    $228.72
    Each
       $228.72
    Supplier Number: 501237
  • Curved
    20 mm
    Log in for availability
    Log in for availability
    Each
    Each - $228.72
    $228.72
    Each
       $228.72
    Supplier Number: 501238