Microelectrode Holders | Avantor
Microelectrode Holders
Microelectrode Holders
Certificates
Microelectrode Holders
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
WPI™s microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers.
  • Female connector
  • Pellet half-cell
  • No pressure Port
  • Screw cap
  • With 6.3 mm Ø
A Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets. The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubber gaskets (without caps).
Filling WPI microelectrode holders with electrolytes containing chloride results in stable electrode potential. Suitable electrolytes include KCl, NaCl and CaCl₂.
Delivery information: Holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. (Call WPI regarding custom designs for other glass diameters.) The holder style you select will depend on your experimental application, space, and instrumentation.
Product Family Options
Informations sur le produit
  • Longueur
    Désignation
    Availability
    Price
  • 27.2 mm
    Microelectrode holder, size: 1.0 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $183.28
    $183.28
    Each
       $183.28
    Supplier Number: MEH3RF10
  • 27.2 mm
    Microelectrode holder, size: 1.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $183.28
    $183.28
    Each
       $183.28
    Supplier Number: MEH3RF15
  • 27.2 mm
    Microelectrode holder, size: 2.0 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $183.28
    $183.28
    Each
       $183.28
    Supplier Number: MEH3RF20