Disposable Scalpels | Avantor
Disposable Scalpels
Disposable Scalpels
Certificates
Disposable Scalpels
7 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The surgical stainless steel blades are installed on plastic handles, and each unit comes sterile and individually wrapped. These are perfect for one time surgical uses, dissections, crafts, student use, woodworking, taxidermy, clay modeling and more.
  • Disposable scalpels perfect for single use, students, crafts and more
  • Stainless steel blade with plastic handle
  • Sterile and individually wrapped
  • Variety of blade styles
  • 10 per box
Each disposable knife comes with a cap to protect the blade for safe disposal or to save the used scalpel for another application. The plastic handle of the scalpel is textured to ensure a firm grip. It is lightweight for comfortable use. And the handle includes a handy, integrated 6 cm metric ruler.
Scalpel blades come in a variety of shapes so that you can choose the ones best suited to your application.
The #10 blade is a commonly used blade for surgical applications. It is used for making large incisions through skin and subcutaneous tissues. It is similar to the #20 blade, which is somewhat larger. The #20 blade is used for larger incisions through thicker tissues or for dissection of the soft tissues in larger animals.
Blades #10, 15, 20, 21, 22 and 23 are all curved and said to have a 'belly'. They are all used for making incisions. The sharpest area on these blades is the apex of curve at the belly. This is the spot on the blade which is used for making the incision. Since the #15 is the smallest of these belly blades, it is often used for small animal surgery where short, precise incisions are required. It could also be used to remove skin lesions, perform an organ biopsy or in fine neurological applications. Larger blades like the #22 and #23 are designed for making bigger incisions like those required for a thoracotomy or a laparotomy.
A #11 blade is a triangular blade with a sharp point, also called a stabbing blade. This scalpel is used for making stab-style incisions and short, precision cuts. Often these cuts are made in shallow or recessed areas. For example, it could be used for inserting chest tubes or drains or for making a cut in a blood vessel where you need to insert a catheter.
The #12 blade is curved and is frequently used for removing sutures. It can be used to hook something that's on a stalk, like a tonsil you wish to remove. Crafters will often select the popular #10 and #11 blades. Surgical blades are typically thinner that craft blades, making them more flexible. Some artists recommend choosing straight blades like the #11 for making straight cuts and curved blades like the #10 for making curved cuts.
Product Family Options
Informations sur le produit
  • Blade No.
    Availability
    Price
  • #10
    Discontinued
    Supplier Number: 500348
  • #11
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - 36,53 $
    36,53 $
    Pack of 10
       36,53 $
    Supplier Number: 500349
  • #12
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - 36,53 $
    36,53 $
    Pack of 10
       36,53 $
    Supplier Number: 500350
  • #15
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - 36,53 $
    36,53 $
    Pack of 10
       36,53 $
    Supplier Number: 500351
  • #20
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - 36,53 $
    36,53 $
    Pack of 10
       36,53 $
    Supplier Number: 500352
  • #21
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - 36,53 $
    36,53 $
    Pack of 10
       36,53 $
    Supplier Number: 500353
  • #22
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - 36,53 $
    36,53 $
    Pack of 10
       36,53 $
    Supplier Number: 500354
  • #23
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - 36,53 $
    36,53 $
    Pack of 10
       36,53 $
    Supplier Number: 500355