Mayo Dissecting Scissors, 14 cm
Mayo Dissecting Scissors, 14 cm
Mayo Dissecting Scissors, 14 cm
Certificates
Mayo Dissecting Scissors, 14 cm
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mayo scissors, also called mayo dissecting scissors, are surgical scissors made popular by the mayo clinic. They have ring handles and long, straight, thick blades with semi-blunt tips that are straight or curved.
  • Blunt, thick blades
  • Ring handles
  • Straight or curved tips
  • Surgical grade stainless steel or premium German steel
The length of the blade determines the reach of the scissors when you are dissecting. The blades are sturdy for heavy duty work like cutting connective tissues, fascia, and sutures.
Mayo scissors are often used for dissection, where the blunt tips prevent accidental damage to surrounding tissue and may be used to delicately probe. They may also be used for dressing and bandage removal.
These surgical scissors are available in surgical grade stainless steel or premium German steel. Both metals are suitable for sterilization by autoclaving or chemically. Always clean your surgical instruments promptly after use. Cleaning helps to minimize staining and preserve the life of your fine surgical instruments.
Product Family Options
Product Information
  • Longueur
    Matière
    Blade Type
    Handle
    Availability
    Price
  • 14 cm (5.5")
    Stainless steel
    Straight
    Ring handle
    Log in for availability
    Log in for availability
    Each
    Each - $99.78
    $99.78
    Each
       $99.78
    Supplier Number: 501749
  • 14 cm (5.5")
    Stainless steel
    Straight
    Ring handle
    Log in for availability
    Log in for availability
    Each
    Each - $86.90
    $86.90
    Each
       $86.90
    Supplier Number: 501749-G
  • 14 cm (5.5")
    Stainless steel
    Curved
    Ring handle
    Discontinued
    Supplier Number: 501750
  • 14 cm (5.5")
    Stainless steel
    Curved
    Ring handle
    Log in for availability
    Log in for availability
    Each
    Each - $86.90
    $86.90
    Each
       $86.90
    Supplier Number: 501750-G