Lockout Stations, Brady®
Supplier:   Avantor
Lockout Stations, Brady®
Supplier:  Avantor
Certificates
Lockout Stations, Brady®
Lockout Stations, Brady®
11 Options Available Below
Product Details & Documents
These stations provide components for locking out different valves, plugs, and electrical and mechanical control points
Standard stations are also sold with safety or steel padlocks and “Do Not Operate” tags made of reusable heavy-duty encapsulated polyester. Each tag also includes a nylon cable tie. Large and extra-large stations also have the additional option of lockable acrylic doors.
Constructed of sturdy white polystyrene with predrilled holes for wall mounting. Stations include peg-board style holes for hooks drilled into back: four for small, six for large, and eight for extra-large. They also feature built-in lock rails (depending on lock size) for up to 15 locks in the small station, up to 20 locks in the large station, and up to 25 locks in the extra-large station.
 : All stations include the following components: one 110 V plug lockout, one 3-in-1 plug lockout, and two labeled lockout hasps. "The small lockout station also includes the following: one mini cable lockout, two 120V snap-on breaker lockouts, two 120/277 V clamp-on breaker lockouts with two cleats, two 480/600 V clamp-on breaker lockouts with two cleats, two universal multi-pole breaker lockouts, one oversized breaker lockout, one wall switch lockout, one small fuse blockout, and one large fuse blockout, and two labeled lockout hasps. Both the large and extra-large lockout stations include one all-purpose cable lockout with 2.4 m (8') sheathed cable, one large universal valve lockout with blocking arm, one 0.6 to 2.5 cm (1/4 to1") ball valve lockout, one 2.5 to 6.4 cm (1 to 21/2") gate valve lockout, and one 2.5–16.5 cm (1 to 61/2") adjustable gate valve lockout. Extra-large lockout stations also include these additional components: two 120 V snap-on breaker lockouts, two 120/277 V clamp-on breaker lockouts with two cleats, two 480/600V clamp-on breaker lockouts with two cleats, two universal multi-pole breaker lockouts, one oversized breaker lockout, one wall switch lockout, one 3.2 to 7.6 cm (11/4 to 3") ball valve lockout, and an extra 2.5 to 6.4 cm (1 to 21/2") gate valve lockout.
Product Family Options
Small Lockout Stations
Informations sur le produit
  • Désignation
    W×D×H
    Availability
    Price
  • With 6 Safety Padlocks and 12 Tags
    30.5 x 10.2 x 45.7 cm (12 x 4 x 18")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 2
    Kit of 2 - 733,78 $
    733,78 $
    Kit of 2
       733,78 $
  • With 6 Steel Padlocks and 12 Tags
    30.5 x 10.2 x 45.7 cm (12 x 4 x 18")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 2
    Kit of 2 - 730,45 $
    730,45 $
    Kit of 2
       730,45 $
Large Lockout Stations
Informations sur le produit
  • Désignation
    W×D×H
    Availability
    Price
  • With 12 Steel Padlocks and 25 Tags
    49.5 x 12.7 x 66 cm (191/2 x 5 x 26")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 2
    Kit of 2 - 1 107,35 $
    1 107,35 $
    Kit of 2
       1 107,35 $
  • Standard
    49.5 x 12.7 x 66 cm (191/2 x 5 x 26")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 2
    Kit of 2 - 711,99 $
    711,99 $
    Kit of 2
       711,99 $
  • With 12 Safety Padlocks and 25 Tags
    49.5 x 12.7 x 66 cm (191/2 x 5 x 26")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 2
    Kit of 2 - 1 011,18 $
    1 011,18 $
    Kit of 2
       1 011,18 $
  • With 12 Safety Padlocks, 25 Tags, and Lockable Acrylic Doors
    49.5 x 12.7 x 66 cm (191/2 x 5 x 26")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 2
    Kit of 2 - 1 096,07 $
    1 096,07 $
    Kit of 2
       1 096,07 $
  • With 12 Steel Padlocks, 25 Tags, and Lockable Acrylic Doors
    49.5 x 12.7 x 66 cm (191/2 x 5 x 26")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 2
    Kit of 2 - 1 085,72 $
    1 085,72 $
    Kit of 2
       1 085,72 $
Extra-Large Lockout Stations
Informations sur le produit
  • Désignation
    W×D×H
    Availability
    Price
  • With 18 Safety Padlocks and 25 Tags
    63.5 x 13.3 x 76.2 cm (25 x 51/4 x 30")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 2
    Kit of 2 - 1 755,54 $
    1 755,54 $
    Kit of 2
       1 755,54 $
  • With 18 Steel Padlocks and 25 Tags
    63.5 x 13.3 x 76.2 cm (25 x 51/4 x 30")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 2
    Kit of 2 - 1 623,92 $
    1 623,92 $
    Kit of 2
       1 623,92 $
  • With 18 Safety Padlocks, 25 Tags, and Lockable Acrylic Doors
    63.5 x 13.3 x 76.2 cm (25 x 51/4 x 30")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 2
    Kit of 2 - 1 789,52 $
    1 789,52 $
    Kit of 2
       1 789,52 $
  • With 18 Steel Padlocks, 25 Tags, and Lockable Acrylic Doors
    63.5 x 13.3 x 76.2 cm (25 x 51/4 x 30")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 2
    Kit of 2 - 1 634,27 $
    1 634,27 $
    Kit of 2
       1 634,27 $