Operating Scissors, Disposable
Operating Scissors, Disposable
Operating Scissors, Disposable
Certificates
Operating Scissors, Disposable
7 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ring-handled operating scissors are surgical scissors commonly used in general surgery for cutting and dissecting tissues. Versatile size that can be used for a variety of procedures, including biological and medical research. These operating scissors are available with straight or curved blades. Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. These surgical scissors are available with a variety of tips, including two sharp pointed blades, one sharp and one blunt blade, or two blunt blades.
  • Great for one-time use or students
  • Straight or curved blades
  • Sharp/sharp, sharp/blunt, or blunt/blunt tips
Our operating scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. These economy scissors are sold in packages of 12, perfect for students or for disposable use. All sales are final on disposable instrument packs.
Product Family Options
Product Information
  • Désignation
    Longueur
    Matière
    Curvature
    Availability
    Price
  • Operating scissors, sharp/blunt
    14 cm (5.5")
    Stainless steel
    Straight
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $71.08
    $71.08
    Pack of 12
       $71.08
    Supplier Number: 503714-12
  • Operating scissors, blunt/blunt
    14 cm (5.5")
    Stainless steel
    Straight
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $71.08
    $71.08
    Pack of 12
       $71.08
    Supplier Number: 503717-12
  • Operating scissors, blunt/blunt
    14 cm (5.5")
    Stainless steel
    Curved
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $71.08
    $71.08
    Pack of 12
       $71.08
    Supplier Number: 503713-12
  • Operating scissors,sharp/sharp
    14 cm (5.5")
    Stainless steel
    Curved
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $71.08
    $71.08
    Pack of 12
       $71.08
    Supplier Number: 503727-12
  • Operating scissors, sharp/blunt
    14 cm (5.5")
    Stainless steel
    Curved
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $71.08
    $71.08
    Pack of 12
       $71.08
    Supplier Number: 503715-12
  • Operating scissors,sharp/sharp
    14 cm (5.5")
    Stainless steel
    Straight
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $71.08
    $71.08
    Pack of 12
       $71.08
    Supplier Number: 503716-12
  • Operating scissors, sharp/blunt
    16 cm (5.5")
    Stainless steel
    Curved
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $71.08
    $71.08
    Pack of 12
       $71.08
    Supplier Number: 503718-12