Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Species:HumanMouseNon-human primateRat
- Description:Collagenase type IV
- Size:1 g
- Cat. no.:MSPP-07427
Spécifications
À propos de cet article
For digestion of native collagen fibrils.
- Maintain receptor integrity with a collagenase preparation low in tryptic activity
- Dissociate pancreatic islet cells effectively
Digest native collagen fibrils in connective tissues with Collagenase Type IV, sourced from Clostridium histolyticum. Collagenase preparations typically contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler and Yaron). Frequently used to dissociate pancreatic islets (Taguchi et al.), and in applications where maintenance of receptor integrity is required, Collagenase Type IV has low tryptic activity compared to other collagenase preparations.