Iris Scissors, Supercut
À propos de cet article
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
- Supercut-one edge micro serrated, one edge honed to the sharpness of a knife
- Sharp tips
11 Options Available Below
Product Details & Documents
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
- Supercut-one edge micro serrated, one edge honed to the sharpness of a knife
- Sharp tips
Product Family Options
Informations sur le produit
- DésignationLongueurMatièreCurvatureAvailabilityPrice
- Iris scissors10 cm (4.0")Stainless steelStraightDiscontinued-Supplier Number: 14218
- Supplier Number: 14218-G
- Supplier Number: 14225-G
- Supplier Number: 503259
- Supplier Number: 14225
- Supplier Number: 503261
- Supplier Number: 14219-G
- Supplier Number: 14219
- Supplier Number: 503260
- Supplier Number: 14224
- Supplier Number: 14224-G
- Supplier Number: 503262