Spring Scissors 4"
Spring Scissors 4"
Spring Scissors 4"
Certificates
Spring Scissors 4"
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Spring scissors are fine surgical instruments for making small, precision cuts. They are 10.5 cm long, and fit comfortably into your hand. The spring mechanism that allows for smooth, precise cutting action, which reduces the risk of tissue damage or trauma. Choose either straight or curved for the 8 mm blade.
  • Spring handles for ambidextrous use
  • Straight or curved tip
  • Surgical grade stainless steel or premium German steel
The curved surgical scissors are often used in ophthalmology to make precise incisions in the cornea or delicate tissues. The curved tip gives greater visibility at the tip of the scissors. The straight blades are more often used in microdissection, microsurgery or plastic surgery. The spring handled surgical scissors are preferred by left-handed surgeons, because they can be used ambidextrously.
This gives the left-handed surgeon a more natural cutting action, which means the surgeon has greater control of the tip and the cutting movement. These are available in surgical grade stainless steel or premium German steel, both of which can withstand high heat for autoclaving or chemical sterilization. To ensure the longevity or your fine surgical scissors, always clean them thoroughly after every use.
Product Family Options
Product Information
  • Désignation
    Longueur
    Matière
    Blade Length
    Curvature
    Availability
    Price
  • Spring scissors
    10.5 cm (4")
    Stainless steel
    8 mm
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $389.30
    $389.30
    Each
       $389.30
    Supplier Number: 501235
  • Spring scissors
    10.5 cm (4")
    Stainless steel
    8 mm
    Curved
    Log in for availability
    Log in for availability
    Each
    Each - $362.55
    $362.55
    Each
       $362.55
    Supplier Number: 14127
  • Spring scissors
    10.5 cm (4")
    German steel
    8 mm
    Curved
    Log in for availability
    Log in for availability
    Each
    Each - $481.92
    $481.92
    Each
       $481.92
    Supplier Number: 14127-G