Swiss Microdissection Scissors, 9 cm
Swiss Microdissection Scissors, 9 cm
Certificates
Swiss Microdissection Scissors, 9 cm
7 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Swiss microdissecting scissors are ring handled surgical scissors particularly suited for dissection and surgical tasks that require precision cutting. These scissors are crafted with high-quality materials and are known for their durability and sharpness. Swiss-made surgical instruments are renowned for their precision and craftsmanship.
  • High quality Swiss-made scissors
  • Sharp, blunt or rounded tips
  • Curved or straight tips
  • Extra fine, fine, or heavy-duty blades
  • Unique beveled design available
  • Swiss surgical grade stainless steel
Swiss manufacturers adhere to strict quality control standards, and their products are known for precision and reliability. WPI Swiss dissecting scissors are made from high-grade stainless steel, which provides corrosion resistance and ensures a long lifespan for the instrument. All these scissors are 9 cm (3.5") long. These scissors have a fine, slender design with sharp blades, allowing for precise cutting in delicate procedures.
They have sharp, blunt or rounded tips These are available with curved or straight tips. You can also get the unique beveled tip design. The handles of Swiss dissecting scissors are designed for ergonomic comfort, providing a secure grip for the surgeon. This is crucial for maintaining control and accuracy during delicate surgical procedures. Like all surgical instruments, Swiss dissecting scissors are designed to be sterilized easily using a chemical sterilization process or by autoclaving.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Sterility
    Availability
    Price
  • Curved, extra fine tips
    Stainless steel
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $138.69
    $138.69
    Each
       $138.69
    Supplier Number: 504519
  • Curved, fine, sharp tips
    Stainless steel
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $148.44
    $148.44
    Each
       $148.44
    Supplier Number: 504520
  • Curved, heavy duty, rounded tips
    Stainless steel
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $146.01
    $146.01
    Each
       $146.01
    Supplier Number: 504521
  • Curved, strong, beveled tips
    Stainless steel
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $150.87
    $150.87
    Each
       $150.87
    Supplier Number: 504522
  • Straight, fine, sharp tips with spear shape
    Stainless steel
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $131.39
    $131.39
    Each
       $131.39
    Supplier Number: 504613
  • Straight, fine, sharp tips
    Stainless steel
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $124.09
    $124.09
    Each
       $124.09
    Supplier Number: 504614
  • Curved, fine blunt tips
    Stainless steel
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $146.01
    $146.01
    Each
       $146.01
    Supplier Number: 504615