Sample Cuvettes
104087-596
: Hach
- Window Material:Optical glass
- Cat. no.:104087-596
- Pathlength:10 mm
Sample cuvettes for photometric measurements.
- Available with or without lid
- Available reusable (made from glass) or for single use (made from plastics)