Iris Scissors, Tungsten Carbide | Avantor
Iris Scissors, Tungsten Carbide
Iris Scissors, Tungsten Carbide
Certificates
Iris Scissors, Tungsten Carbide
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
  • Tungsten carbide insert
  • Stainless steel
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Availability
    Price
  • Iris scissor, straight
    11 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $216.55
    $216.55
    Each
       $216.55
    Supplier Number: 500216
  • Iris scissor, straight, german
    11 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $235.42
    $235.42
    Each
       $235.42
    Supplier Number: 500216-G
  • Iris scissor, curved
    11 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $221.42
    $221.42
    Each
       $221.42
    Supplier Number: 500217
  • Iris scissor, curved, german
    11 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $235.42
    $235.42
    Each
       $235.42
    Supplier Number: 500217-G