Tenotomy Scissors
Tenotomy Scissors
Certificates
Tenotomy Scissors
14 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Tenotomy scissors are used for delicate procedures. With the long handle and thin tapered blades, the tenotomy scissors are able to cut tissues in hard to reach, limited spaces.
  • One serrated blade, one honed razor sharp
  • Sharp tips
  • Surgical grade stainless steel
These sharp, straight scissors make small, precision cuts, and they are perfect for delicate operations. The longer handles help to control the precision of these tenotomy scissors in restricted spaces or with delicate membranes or tissues, like brain tissue or ophthalmic surgeries.
These scissors are designed for probing and cutting in tight spots. A precision pair of scissors like the tenotomy scissors limits the damage to surrounding tissues, which is critical in neurosurgery and other applications.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Matière
    Availability
    Price
  • Supercut scissor, straight
    10 cm
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $194.67
    $194.67
    Each
       $194.67
    Supplier Number: 14395
  • Supercut scissor, curved
    10 cm
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $201.97
    $201.97
    Each
       $201.97
    Supplier Number: 14396
  • Supercut scissor, curved, german
    10 cm
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $168.60
    $168.60
    Each
       $168.60
    Supplier Number: 14396-G
  • Supercut scissor, straight, german
    11.5 cm
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $188.42
    $188.42
    Each
       $188.42
    Supplier Number: 14395-G
  • Supercut scissor, straight
    11.5 cm
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $194.67
    $194.67
    Each
       $194.67
    Supplier Number: 503263
  • Supercut scissor, curved
    11.5 cm
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $194.67
    $194.67
    Each
       $194.67
    Supplier Number: 503264
  • Supercut scissor, straight
    12.5 cm
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $231.15
    $231.15
    Each
       $231.15
    Supplier Number: 14220
  • Supercut scissor, straight, german
    12.5 cm
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $170.66
    $170.66
    Each
       $170.66
    Supplier Number: 14220-G
  • Supercut scissor, curved
    12.5 cm
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $226.29
    $226.29
    Each
       $226.29
    Supplier Number: 14221
  • Supercut scissor, curved, german
    12.5 cm
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $195.02
    $195.02
    Each
       $195.02
    Supplier Number: 14221-G
  • Supercut scissor, straight
    14.0 cm
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $226.29
    $226.29
    Each
       $226.29
    Supplier Number: 503265
  • Supercut scissor, curved
    14.0 cm
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $214.12
    $214.12
    Each
       $214.12
    Supplier Number: 503266
  • Supercut scissor, straight
    16.0 cm
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $236.04
    $236.04
    Each
       $236.04
    Supplier Number: 503267
  • Supercut scissor, curved
    16.0 cm
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $248.17
    $248.17
    Each
       $248.17
    Supplier Number: 503268