Metzenbaum Scissors
Metzenbaum Scissors
Metzenbaum Scissors
Certificates
Metzenbaum Scissors
11 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue.
The tips are semi-blunt, and they are available in curved or straight styles.
Product Family Options
Product Information
  • Désignation
    Longueur
    Matière
    Availability
    Price
  • Metzenbaum scissors, straight tips
    11.5 cm (4.5")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $94.91
    $94.91
    Each
       $94.91
    Supplier Number: 501747
  • Metzenbaum scissors, straight tips
    11.5 cm (4.5")
    German steel
    Log in for availability
    Log in for availability
    Each
    Each - $86.90
    $86.90
    Each
       $86.90
    Supplier Number: 501747-G
  • Metzenbaum scissors, curved tips
    11.5 cm (4.5")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $94.91
    $94.91
    Each
       $94.91
    Supplier Number: 501748
  • Metzenbaum scissors, curved tips
    11.5 cm (4.5")
    German steel
    Log in for availability
    Log in for availability
    Each
    Each - $86.90
    $86.90
    Each
       $86.90
    Supplier Number: 501748-G
  • Metzenbaum scissors, straight tips
    14.5 cm (5.75")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $99.78
    $99.78
    Each
       $99.78
    Supplier Number: 501252
  • Metzenbaum scissors, straight tips
    14.5 cm (5.75")
    German steel
    Log in for availability
    Log in for availability
    Each
    Each - $86.90
    $86.90
    Each
       $86.90
    Supplier Number: 501252-G
  • Metzenbaum scissors, curved tips
    14.5 cm (5.75")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $99.78
    $99.78
    Each
       $99.78
    Supplier Number: 501253
  • Metzenbaum scissors, curved tips
    14.5 cm (5.75")
    German steel
    Log in for availability
    Log in for availability
    Each
    Each - $86.90
    $86.90
    Each
       $86.90
    Supplier Number: 501253-G
  • Metzenbaum scissors, straight tips
    17.5 cm (7")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $99.78
    $99.78
    Each
       $99.78
    Supplier Number: 501254
  • Metzenbaum scissors, straight tips
    17.5 cm (7")
    German steel
    Log in for availability
    Log in for availability
    Each
    Each - $83.74
    $83.74
    Each
       $83.74
    Supplier Number: 501254-G
  • Metzenbaum scissors, curved tips
    17.5 cm (7")
    Stainless steel
    Log in for availability
    Log in for availability
    Each
    Each - $99.78
    $99.78
    Each
       $99.78
    Supplier Number: 501255