Notrax® 765 Deep Freeze Mat
Fournisseur: NOTRAX USA, INC.
New Product
Certificats
À propos de cet article
The NoTrax Deep Freeze Mat™ 765 provides the ultimate solution for keeping your freezer floor clean, safe, and slip-free. This innovative freezer mat is engineered to withstand extreme cold temperatures while providing exceptional performance and durability.
- Made from a tough rubber compound
- Raised knob solid top surface that enhances traction
- Tested to withstand temperatures to −40 °F (approximately −40 °C)
- Manufactured with Microstop anti-microbial rubber compound that inhibits growth of bacteria and fungi
- Low profile allows for easy on/off cart traffic
- High quality walk-in deep freezer mat
This anti-slip mat features a raised knob solid top surface to enhance traction, and is designed for use in meat lockers, walk-ins, and other specialty freezer applications. Manufactured with Microstop anti-microbial rubber compound.
Caution: This product can expose you to chemicals including disodecyl phthalate which is known to the State of California to cause birth defects or reproductive harm.
Frequently Bought Together