Filling Solutions
Filling Solutions
Filling Solutions
Filling solutions to use with ISO-NOP, ISO-OXY-2 or OXELP oxygen sensors.
Product Family Options
  • Désignation
    Taille
    Fill Solution
  • Filling solution
    10 ml
    Nitric oxide
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $160.61
    $160.61
    Each(1/Itm/Bx)
       $160.61
     : 7325
  • Filling solution
    10 ml
    Oxygen
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $158.17
    $158.17
    Each(1/Itm/Bx)
       $158.17
     : 7326