Weitlaner Retractors
Weitlaner Retractors
Weitlaner Retractors
Certificates
Weitlaner Retractors
7 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made up of stainless steel.
Product Family Options
Self-retaining (A)
Product Information
  • Désignation
    Longueur
    Availability
    Price
  • Weitlaner retractor, self-retaining, 3×4 prongs, 6.5 cm maximum spread, blunt
    16.5 cm (6.5")
    Log in for availability
    Log in for availability
    Each
    Each - $413.61
    $413.61
    Each
       $413.61
    Supplier Number: 501724
  • Weitlaner retractor, self-retaining, 3×4 prongs, 6.5 cm maximum spread, blunt, German
    16.5 cm (6.5")
    Log in for availability
    Log in for availability
    Each
    Each - $357.06
    $357.06
    Each
       $357.06
    Supplier Number: 501724-G
  • Weitlaner retractor, self-retaining, 3×4 prongs, 7 cm maximum spread, sharp
    16.5 cm (6.5")
    Log in for availability
    Log in for availability
    Each
    Each - $413.61
    $413.61
    Each
       $413.61
    Supplier Number: 501723
  • Weitlaner retractor, self-retaining, 3×4 prongs, 7 cm maximum spread, sharp, German
    16.5 cm (6.5")
    Log in for availability
    Log in for availability
    Each
    Each - $357.06
    $357.06
    Each
       $357.06
    Supplier Number: 501723-G
Self-retaining (B)
Product Information
  • Désignation
    Longueur
    Availability
    Price
  • Weitlaner retractor, 3×4 prongs, 4 cm, maximum spread, sharp
    14 cm (5.5")
    Log in for availability
    Log in for availability
    Each
    Each - $335.77
    $335.77
    Each
       $335.77
    Supplier Number: 501721
  • Weitlaner retractor, self-retaining, 3×4 prongs, 4 cm maximum spread, sharp, German
    14 cm (5.5")
    Log in for availability
    Log in for availability
    Each
    Each - $428.20
    $428.20
    Each
       $428.20
    Supplier Number: 501721-G
  • Weitlaner retractor, 3×4 prongs, 4.5 cm maximum spread, blunt
    14 cm (5.5")
    Log in for availability
    Log in for availability
    Each
    Each - $389.30
    $389.30
    Each
       $389.30
    Supplier Number: 501722