Fine Mesh Baskets with Lids
À propos de cet article
Made up of stainless steel 304.
- Electropolished
- Burr free to protect wraps and prevent injury
- Smooth rounded corners
- Whole and wires size: 1.6×1.6×0.7 mm
- Top and lid frame wire diameter: 3.5 and 4.0 mm (depending on the basket)
- Fine mesh with opening squares of 1.6 mm and wire diameter of 0.7 mm
- Wire locking clip
Applications:
Micro surgical instrument cleaning and sterilization
Micro surgical instrument storage
27 Options Available Below
Product Details & Documents
Made up of stainless steel 304.
- Electropolished
- Burr free to protect wraps and prevent injury
- Smooth rounded corners
- Whole and wires size: 1.6×1.6×0.7 mm
- Top and lid frame wire diameter: 3.5 and 4.0 mm (depending on the basket)
- Fine mesh with opening squares of 1.6 mm and wire diameter of 0.7 mm
- Wire locking clip
Applications:
Micro surgical instrument cleaning and sterilization
Micro surgical instrument storage
Product Family Options
Informations sur le produit
- DimensionsAvailabilityPrice
- Supplier Number: WP-4344Spécifications:
More
Specifications
Cat. No.77665-782 - Supplier Number: WP-4345Spécifications:
More
Specifications
Cat. No.77665-786 - Supplier Number: WP-4346Spécifications:
More
Specifications
Cat. No.77665-790 - Supplier Number: WP-4348Spécifications:
More
Specifications
Cat. No.77665-796 - Supplier Number: WP-4349Spécifications:
More
Specifications
Cat. No.77665-800 - Supplier Number: WP-4353Spécifications:
More
Specifications
Cat. No.77665-814 - Supplier Number: WP-4354Spécifications:
More
Specifications
Cat. No.77665-818 - Supplier Number: WP-4350Spécifications:
More
Specifications
Cat. No.77665-804 - Supplier Number: WP-4351Spécifications:
More
Specifications
Cat. No.77665-808 - Supplier Number: WP-4355Spécifications:
More
Specifications
Cat. No.77665-822 - Supplier Number: WP-4356Spécifications:
More
Specifications
Cat. No.77665-826 - Supplier Number: WP-4357Spécifications:
More
Specifications
Cat. No.77665-828 - Supplier Number: WP-4367Spécifications:
More
Specifications
Cat. No.77665-848 - Supplier Number: WP-4359Spécifications:
More
Specifications
Cat. No.77665-832 - Supplier Number: WP-4358Spécifications:
More
Specifications
Cat. No.77665-830 - Supplier Number: WP-4368Spécifications:
More
Specifications
Cat. No.77665-850 - Supplier Number: WP-4369Spécifications:
More
Specifications
Cat. No.77665-852 - Supplier Number: WP-4360Spécifications:
More
Specifications
Cat. No.77665-834 - Supplier Number: WP-4361Spécifications:
More
Specifications
Cat. No.77665-836 - Supplier Number: WP-4362Spécifications:
More
Specifications
Cat. No.77665-838 - Supplier Number: WP-4365Spécifications:
More
Specifications
Cat. No.77665-844 - Supplier Number: WP-4370Spécifications:
More
Specifications
Cat. No.77665-854 - Supplier Number: WP-4363Spécifications:
More
Specifications
Cat. No.77665-840 - Supplier Number: WP-4371Spécifications:
More
Specifications
Cat. No.77665-856 - Supplier Number: WP-4364Spécifications:
More
Specifications
Cat. No.77665-842 - Supplier Number: WP-4366Spécifications:
More
Specifications
Cat. No.77665-846 - Supplier Number: WP-4372Spécifications:
More
Specifications
Cat. No.77665-858