Respirator Cleaning Wipes, 3M™
Catalogue # CA56616-231
Fournisseur: 3M
Spécifications
- Description:Respirator cleaning wipes
- Cat. no.:CA56616-231
- Supplier No.:504
Spécifications
À propos de cet article
Ideal for cleaning elastomeric facepiece respirators.
- Individually packed, formulated with alcohol-free fragrance, will not damage facepiece materials
- Dimensions: 20.3×25.4 cm (8×10")
Packaging: Packaged in dispenser box.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.