Model MI-150 Microscopy Illuminator, Dolan-Jenner | Avantor
Model MI-150 Microscopy Illuminator, Dolan-Jenner
Supplier:   Dolan-Jenner
Model MI-150 Microscopy Illuminator, Dolan-Jenner
Supplier:  Dolan-Jenner
Certificates
Model MI-150 Microscopy Illuminator, Dolan-Jenner
3 Options Available Below
Product Details & Documents
The MI-150 is a 150-watt quartz halogen illuminator designed specifically for laboratory use
  • Colour temperature: 3200 °K
  • Lamp: EKE, 21 V, 150 W (200 hours at full intensity)
Optional versions of the MI-150 come with a standard A3739P ringlight (41446-978), with BG2820 single gooseneck self-supporting cable and one LH-759 lens (41446-974), or with EEG2823 dual gooseneck self-supporting cable and two LH-759 lenses (41446-976).
The lamp delivers nearly 400,000 foot-candles of high-intensity, cold illumination with minimal housing vibration or noise. The illuminator features a sleek plastic housing, minimal light leakage, and 0-100% solid state intensity control.
Certifications: UL listed. CE mark.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 41446-974
  • 41446-976
  • 41446-978
  • 41446-970
  • Dimensions:
  • 22.2L x 15.24W x 14H cm (83/4 x 6 x 51/2")
  • 22.2L x 15.24W x 14H cm (83/4 x 6 x 51/2")
  • 22.2L x 15.24W x 14H cm (83/4 x 6 x 51/2")
  • 22.2L x 15.24W x 14H cm (83/4 x 6 x 51/2")
  • Electrical:
  • 110/120 or 220/230V AC, 50/60 Hz
  • 110/120 or 220/230V AC, 50/60 Hz
  • 110/120 or 220/230V AC, 50/60 Hz
  • 110/120 or 220/230V AC, 50/60 Hz
  • Weight:
  • 3.5 kg (7.75 lbs.)
  • 3.5 kg (7.75 lbs.)
  • 3.5 kg (7.75 lbs.)
  • 3.5 kg (7.75 lbs.)
Spécifications
Product Family Options
Informations sur le produit
  • Modèle
    Availability
    Price
  • Illuminator, Single Gooseneck
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,536.54
    $2,536.54
    Each
       $2,536.54
    Supplier Number: MI-150SG
    Spécifications:
    UL Listed
    More
    Specifications
    Cat. No.
    41446-974
    Dimensions
    22.2L x 15.24W x 14H cm (83/4 x 6 x 51/2")
    Electrical
    110/120 or 220/230V AC, 50/60 Hz
    Weight
    3.5 kg (7.75 lbs.)
    UL Listed
  • Illuminator, Dual Gooseneck
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,478.44
    $3,478.44
    Each
       $3,478.44
    Supplier Number: MI-150DG
    Spécifications:
    UL Listed
    More
    Specifications
    Cat. No.
    41446-976
    Dimensions
    22.2L x 15.24W x 14H cm (83/4 x 6 x 51/2")
    Electrical
    110/120 or 220/230V AC, 50/60 Hz
    Weight
    3.5 kg (7.75 lbs.)
    UL Listed
  • Illuminator with Ringlight
    Discontinued
    Supplier Number: -
    Spécifications:
    UL Listed
    More
    Specifications
    Cat. No.
    41446-978
    Dimensions
    22.2L x 15.24W x 14H cm (83/4 x 6 x 51/2")
    Electrical
    110/120 or 220/230V AC, 50/60 Hz
    Weight
    3.5 kg (7.75 lbs.)
    UL Listed
  • Illuminator
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,753.61
    $1,753.61
    Each
       $1,753.61
    Supplier Number: MI-150
    Spécifications:
    UL Listed
    More
    Specifications
    Cat. No.
    41446-970
    Dimensions
    22.2L x 15.24W x 14H cm (83/4 x 6 x 51/2")
    Electrical
    110/120 or 220/230V AC, 50/60 Hz
    Weight
    3.5 kg (7.75 lbs.)
    UL Listed