HNE Buffer, Sterile
HNE Buffer, Sterile
1000 ml, 1 Bottle per pack
Catalog # CA77623-688
Supplier:   Teknova
Supplier #:  21509
HNE Buffer, Sterile
Catalog # CA77623-688
Supplier:  Teknova
CAS Number:  
HNE Buffer, Sterile
1000 ml, 1 Bottle per pack
CAS Number:  
Price
$155.92
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
HNE Buffer composition: 50 mM HEPES, 0.15 M sodium chloride, 25 mM EDTA.
Specifications
Specifications
  • Type de conditionnement:
    1 Bottle per pack
  • Taille:
    1000 ml
  • Cat. No.:
    CA77623-688
  • Storage temperature:
    Store at room temperature
  • Shelf life:
    1095 days