Vannas Spring Scissors | Avantor
Vannas Spring Scissors
Vannas Spring Scissors
Certificates
Vannas Spring Scissors
7 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Vannas scissors are specialized spring scissors designed for precision cutting and often used by scientific researchers.
  • Stainless steel and Premium German steel
They are named after the Swedish scientist Gunnar Vannas who invented them in the 1940s. Compared with other spring scissors, vannas scissors are distinguished by their delicate, thin, sharp tips. The slender, pointed tips allows for precise cutting with minimal damage to surrounding tissues. Manufactured with surgical grade stainless steel, these scissors are durable and resistant to corrosion.
Vannas scissors are popular choice for researchers in fields that require precision cutting, such as biology, neuroscience, and microsurgery.
Common applications include:
Microdissection of small organisms, such as insects or embryos
Preparation of small tissue samples for histological analysis
Cutting thin sections of cells/tissues for viewing under a microscope
Preparation of brain slices for electrophysiological studies
Dissection of small neural circuits
Preparation of brain tissue for imaging or analysis
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Matière
    Blade Length
    Curvature
    Tip Size
    Availability
    Price
  • Vannas spring scissors
    8 cm (3.5")
    Stainless steel
    5 mm
    Straight
    0.1 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $507.20
    $507.20
    Each
       $507.20
    Supplier Number: 14003
  • Vannas spring scissors
    8 cm (3.5")
    German steel
    5 mm
    Straight
    0.1 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $578.28
    $578.28
    Each
       $578.28
    Supplier Number: 14003-G
  • Vannas spring scissors
    8 cm (3.5")
    Stainless steel
    5 mm
    Curved
    0.1 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $690.97
    $690.97
    Each
       $690.97
    Supplier Number: 14122
  • Vannas spring scissors
    8 cm (3.5")
    German steel
    5 mm
    Curved
    0.1 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $589.34
    $589.34
    Each
       $589.34
    Supplier Number: 14122-G
  • Vannas spring scissors
    8 cm (3.5")
    Stainless steel
    5 mm
    Angled
    0.1 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $669.08
    $669.08
    Each
       $669.08
    Supplier Number: 500260
  • Vannas spring scissors
    8 cm (3.5")
    German steel
    5 mm
    Angled
    0.1 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $587.76
    $587.76
    Each
       $587.76
    Supplier Number: 500260-G
  • Vannas spring scissors
    8 cm (3.5")
    Stainless steel
    5 mm
    Angled
    0.1 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $647.20
    $647.20
    Each
       $647.20
    Supplier Number: 501790