Vessel Clips
Vessel Clips
Vessel Clips
Certificates
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Vessel clips are a surgical instrument used to clamp blood vessels during surgical procedures to temporarily occlude (block) blood flow in a specific area. These clips are made of metal and come in a variety of sizes. These ligating vessel clips can tie off blood vessels or other tissues.
  • Variety of tip sizes
  • Surgical grade stainless steel
The size of the vessel clip you need can vary depending on the size of the vessel or tissue being occluded. These are available with 0.8×5 mm, 1×6 mm, 1.5×5 mm and 1.5×8 mm jaws. These vessel clips are manufactured from surgical grade stainless steel, and they are suitable for autoclaving or chemical sterilization. Always clean surgical instruments thoroughly and promptly.
Packaging: They come in packages of five clips.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Vessel clip, 0.8×5 mm jaws
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $182.49
    $182.49
    Pack of 5
       $182.49
    Supplier Number: 15911
  • Vessel clip, 1×6 mm jaws
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $182.49
    $182.49
    Pack of 5
       $182.49
    Supplier Number: 15913
  • Vessel clip, 1.5×5 mm jaws
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $257.92
    $257.92
    Pack of 5
       $257.92
    Supplier Number: 14120
  • Vessel clip, 1.5×8 mm jaws
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $257.92
    $257.92
    Pack of 5
       $257.92
    Supplier Number: 14121