VWR® Traceable® Manometers
VWR® Traceable® Manometers
VWR® Traceable® Manometers
VWR® Traceable® Manometers
These manometers display gauge and differential pressure or vacuum readings in eleven different units.
  • Traceable®
  • Backlit for low-light conditions
  • Memory feature recalls highest and lowest readings
  • Automatic capture of data
Differential feature allows reading the difference of the pressure or vacuum between two different sources
Count-up timer displays up to 99 hours, 59 minutes, 59 seconds. Up to 99 readings can be manually captured and printed. Data logging feature allows automatic capture and storage of 12,000 readings. Printer output includes samples number, current reading, date, and time.
A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
LCD digits are 1.6 cm (5/8") high. Brilliant backlighting allows reading in dark areas. Hose fittings accept hose or tubing with inside diameters from 0.2 to 0.5 cm (1/16 to 3/16"). Accuracy: ±0.39% of full scale between 0 to 50°C (32 to 122°F).
  • Catalog No:
  • 89094-722
  • 89094-728
  • Model:
  • 89094-722
  • 89094-728
  • Range:
  • –345 to 345 mbar, –0.345 to 0.345 bar, –351 to 351cm/H2O, –138 to 138 in./H2O, –12 to 12 ft./H2O, –259 to 259 mm/Hg, –10 to 10 in./Hg, –0.35 to 0.35 kg/cm2, –80 to 80 oz./in.2, –34.5 to 34.5 kPa
  • –6895 to 6895 mbar, –6.895 to 6.895 bar, –7031 to 7031cm/H2O, –2768 to 2768 in./H2O, –231 to 231 ft./H2O, –5171 to 5171 mm/Hg, –204 to 204 in./Hg, –7.03 to 7.03 kg/cm2, –1600 to 1600 oz./in.2, –690 to 690 kPa
  • Max. pressure:
  • 20 psi
  • 150 psi
  • Resolution:
  • 0.003 psi, 0.2 mbar, 0.001 bar, 0.2 cm/H2O, 0.1 in./H2O, 0.01 ft./H2O, 0.2 mm/Hg, 0.01 in./Hg, 0.001kg/cm2, 0.05 oz./in.2, 0.02 kPa
  • 0.1 psi, 4 mbar, 0.004 bar, 4 cm/H2O, 2 in./H2O, 0.2 ft./H2O, 3 mm/Hg, 1 in./Hg, 0.004 kg/cm2, 1oz./in.2, 0.4 kPa
Product Family Options
Meters
  • Pressure Range
  • −5 - 5 psig
    Each
    Each - $1,619.89
    $1,619.89
    Each
       $1,619.89
    Specifications
    Cat. No.
    89094-722
    Model
    89094-722
    Range
    –345 to 345 mbar, –0.345 to 0.345 bar, –351 to 351cm/H2O, –138 to 138 in./H2O, –12 to 12 ft./H2O, –259 to 259 mm/Hg, –10 to 10 in./Hg, –0.35 to 0.35 kg/cm2, –80 to 80 oz./in.2, –34.5 to 34.5 kPa
    Max. pressure
    20 psi
    Resolution
    0.003 psi, 0.2 mbar, 0.001 bar, 0.2 cm/H2O, 0.1 in./H2O, 0.01 ft./H2O, 0.2 mm/Hg, 0.01 in./Hg, 0.001kg/cm2, 0.05 oz./in.2, 0.02 kPa
  • −100 - 100 psig
    Each
    Each - $1,246.06
    $1,246.06
    Each
       $1,246.06
    Specifications
    Cat. No.
    89094-728
    Model
    89094-728
    Range
    –6895 to 6895 mbar, –6.895 to 6.895 bar, –7031 to 7031cm/H2O, –2768 to 2768 in./H2O, –231 to 231 ft./H2O, –5171 to 5171 mm/Hg, –204 to 204 in./Hg, –7.03 to 7.03 kg/cm2, –1600 to 1600 oz./in.2, –690 to 690 kPa
    Max. pressure
    150 psi
    Resolution
    0.1 psi, 4 mbar, 0.004 bar, 4 cm/H2O, 2 in./H2O, 0.2 ft./H2O, 3 mm/Hg, 1 in./Hg, 0.004 kg/cm2, 1oz./in.2, 0.4 kPa