ContactUsLinkComponent
Analytical Grinding Mill, A11, IKA® Works
Analytical Grinding Mill, A11, IKA® Works
Supplier:  IKA Works
Certificates
undefined
Analytical Grinding Mill, A11, IKA® Works
Supplier:  IKA Works
Specifications
  • Catalog No:
  • 33998-410
  • 33998-408
  • Dimensions:
  • 8.5W x 8.5D x 24H cm (33/8 x 33/8 x 91/2")
  • 8.5W x 8.5D x 24H cm (33/8 x 33/8 x 91/2")
  • Max. speed:
  • 28,000 rpm
  • 28,000 rpm
  • Unit weight:
  • 1.5 kg (3.3 lbs.)
  • 1.5 kg (3.3 lbs.)