Spring Scissors, Round Handles
Spring Scissors, Round Handles
Spring Scissors, Round Handles
Certificates
Spring Scissors, Round Handles
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Round handle spring scissors have a rounded, ergonomic handle design and a spring mechanism that allows for smooth, precise cutting action. The rounded handle design offers a comfortable and natural grip, reducing hand fatigue and improving control during use. This can be especially beneficial for long procedures that require a lot of cutting. The spring mechanism provides a smooth, precise cutting action, which reduces the risk of tissue damage or trauma.
  • 14 cm (5.5") long
  • 6.5 mm blades
  • Stainless steel
The 6.5 mm curved blade is often used in ophthalmology to make precise incisions in the cornea or delicate tissues. The straight blades are more often used in microdissection, microsurgery or plastic surgery. These stainless steel spring scissors are durable and may be sterilized by chemical sterilant or autoclaving.
Product Family Options
Product Information
  • Blade Length
    Blade Type
    Handle
    Availability
    Price
  • 6.5 mm
    Straight
    Round
    Log in for availability
    Log in for availability
    Each
    Each - $335.77
    $335.77
    Each
       $335.77
    Supplier Number: 14111
  • 6.5 mm
    Straight german
    Round
    Log in for availability
    Log in for availability
    Each
    Each - $314.42
    $314.42
    Each
       $314.42
    Supplier Number: 14111-G
  • 7 mm
    Curved
    Round
    Log in for availability
    Log in for availability
    Each
    Each - $345.50
    $345.50
    Each
       $345.50
    Supplier Number: 14112