Sterilisation Baskets, Side Perforated, Flat Base
À propos de cet article
Stainless steel sterilization baskets for instrument cleaning, sterilization and storage. The perforated side walls protect instrument edges and tips from damage. These sterilization baskets feature electropolished smooth, rounded corners. The baskets are burr free to protect wraps and prevent injury.
28 Options Available Below
Product Details & Documents
Stainless steel sterilization baskets for instrument cleaning, sterilization and storage. The perforated side walls protect instrument edges and tips from damage. These sterilization baskets feature electropolished smooth, rounded corners. The baskets are burr free to protect wraps and prevent injury.
Product Family Options
Informations sur le produit
- MatièreDimensionsAvailabilityPrice
- Supplier Number: WP-4545
- Supplier Number: WP-4546
- Supplier Number: WP-4547
- Supplier Number: WP-4548
- Supplier Number: WP-4549
- Supplier Number: WP-4550
- Supplier Number: WP-4551
- Supplier Number: WP-4552
- Supplier Number: WP-4553
- Supplier Number: WP-4554
- Supplier Number: WP-4555
- Supplier Number: WP-4556
- Supplier Number: WP-4557
- Supplier Number: WP-4558
- Supplier Number: WP-4559
- Supplier Number: WP-4560
- Supplier Number: WP-4561
- Supplier Number: WP-4562
- Supplier Number: WP-4563
- Supplier Number: WP-4564
- Supplier Number: WP-4565
- Supplier Number: WP-4566
- Supplier Number: WP-4567
- Supplier Number: WP-4568
- Supplier Number: WP-4569
- Supplier Number: WP-4570
- Supplier Number: WP-4571
- Supplier Number: WP-4572