Standard Wire Mesh Sterilization Baskets, with Drop Handles
Supplier: WORLD PRECISION INSTRUMENTS LLC
New Product
Certificates
About this item
Electropolished, burr free, and stackable wire mesh sterilization basket with drop handles for instrument cleaning, sterilizatition and storage.
- Manufactured with stainless steel 304
- Electropolished
- Burr free to protect wraps and prevent injury
- Smooth rounded corners
- Baskets are stackable
- Handles are mounted on the inside and drop inside the basket for stacking
Meets all the requirements of automatic washing and sterilization for all types of instruments.
Top and bottom frame diameter: 5.0 mm
Wire mesh with mesh openings of 4.5 mm, wire diameter of 1.0 mm
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Single Frame Lids
Product Information
Frequently Bought Together