Planet-Safe® Sampling Spoons | Avantor
Planet-Safe® Sampling Spoons
Supplier:   Diversified Biotech
Planet-Safe® Sampling Spoons
Certificates
Planet-Safe® Sampling Spoons
2 Options Available Below
Product Details & Documents
Made of PLA, a 100% planet-based bio-polymer, sustainabley derived from renewable resources.
  • Proprietary formulation won’t warp in boiling liquids
  • 10 ml level capacity
  • Individually wrapped and sterile
Sit-flat foot keeps spoons level during weighing. Rigid construction allows easy break up of caked powders. Safe to incinerate and commercially compostable.
Product Family Options
Informations sur le produit
  • Longueur
    Color
    Capacité
    Availability
    Price
  • 4"
    Green
    10 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25 (10ml)
    Pack of 25 (10ml) - $52.58
    $52.58
    Pack of 25 (10ml)
       $52.58
    La quantité minimale de commande est de 1
    Supplier Number: BPSP-1000
  • 6"
    Green
    10 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25 (10ml)
    Pack of 25 (10ml) - $52.58
    $52.58
    Pack of 25 (10ml)
       $52.58
    La quantité minimale de commande est de 1
    Supplier Number: BPSP-2000