SHUR/Sharp™ Microtome Blades, Disposable, TBS® | Avantor
SHUR/Sharp™ Microtome Blades, Disposable, TBS®
SHUR/Sharp™ Microtome Blades, Disposable, TBS®
Certificates
SHUR/Sharp™ Microtome Blades, Disposable, TBS®
SHUR/Sharp™ Microtome Blades, Disposable, TBS®
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
SHURSharp™ offers three disposable microtome blades - a Teflon coated high-profile blade, a low-profile blade and heavy duty blade.
  • Proprietary coating process minimizes compression and produces excellent ribbon
  • SHURSharp™ low or high profile blades provide consistent sharpness and dependability
Blades are intended for use with the SHURSharp™ microtome blade holders. Low- and high-profile blades can also be used with most low/high-profile blade holders.
High-profile and low-profile blades will fit into any corresponding high or low profile blade holders. Heavy-duty blade fits only in TBS blade holder for cutting denser specimens.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Low-Profile Blades, Teflon Coated, Brown Package
    Discontinued
    Supplier Number: -
  • High-Profile Blades, Teflon Coated, Green Package
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $738.08
    $738.08
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $7,380.80
    $7,380.80
    Pack of 50
       $738.08
    Case of 500
       $7,380.80
    Supplier Number: DMB-HP
  • Heavy-Duty Blades, Uncoated, Requires DMBH or DMBH-E, Blue Package
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 35
    Pack of 35 - $878.09
    $878.09
    Log in pour la disponibilité
    Case of 350
    Case of 350 - $8,780.90
    $8,780.90
    Pack of 35
       $878.09
    Case of 350
       $8,780.90
    Supplier Number: DMB-HD