VWR® High-Temperature Blast Burner
VWR® high-temperature blast burner
Catalog # 89038-548
Supplier: VWR International
Jump to:
Product Details & Documents
Designed to burn all gases in large volume with short, wide, uniform flame.
- Safety cap
- Short, wide, uniform flame
- Micrometer needle valve
Special micrometer needle valve regulates gas. Safety cap prevents dangerous backflashes. Injector-shaped 2.9mm (1/8") venturi tube. BTU output: 3800. Inlet O.D.: 10mm (13/32"). Overall height: 20.3 cm (8").
Specifications
- Désignation:VWR® high-temperature blast burner