Four-Circle Perf Sheets
Four-circle perf sheets, 12×7"
Catalog # 77661-498
Supplier: REVVITY HEALTH SCIENCES, INC.
Supplier #: GR2266004
Jump to:
Product Details & Documents
Revvity 226 filter paper sheets for component of medical device.
- Ahlstrom 226 filter paper is 100% cotton linter fiber filter paper
100% pure cotton linter filter paper provides even and uniform sample distribution. This feature delivers reliable and homogenous analytical results from across the dried sample area. Simply apply your sample, allow it to air dry completely, and then you are ready to ship or analyze.
Certifications: Components are not certified.
Packaging: Sold in packs of 100 sheets.
Specifications
- Désignation:Four-circle perf sheets
- Dimensions:12×7"