Three-Circle Perf Sheets
Three-circle perf sheets, 12×13"
Catalog # 77661-496
Supplier: REVVITY HEALTH SCIENCES, INC.
Fournisseur #: GR2266003
Jump to:
Product Details & Documents
Revvity 226 filter paper sheets for component of medical device.
- Ahlstrom 226 filter paper is 100% cotton linter fiber filter paper
100% pure cotton linter filter paper provides even and uniform sample distribution. This feature delivers reliable and homogenous analytical results from across the dried sample area. Simply apply your sample, allow it to air dry completely, and then you are ready to ship or analyze.
Certifications: Components are not certified.
Packaging: Sold in packs of 100 sheets.
Spécifications
- Désignation:Three-circle perf sheets
- Dimensions:12×13"