Grade 226 Sheets, 8×10"
Grade 226 sheets, 8×10"
Catalog # 77661-492
Supplier: REVVITY HEALTH SCIENCES, INC.
Supplier #: GR2266001
Jump to:
Product Details & Documents
Revvity 226 filter paper sheets for component of medical device, blank device component.
- Ahlstrom 226 filter paper is 100% cotton linter fiber filter paper
- Blank, no printing
100% pure cotton linter filter paper provides even and uniform sample distribution. This feature delivers reliable and homogenous analytical results from across the dried sample area. Simply apply your sample, allow it to air dry completely, and then you are ready to ship or analyze.
Certifications: Components are not certified.
Packaging: Sold in packs of 100 sheets.
Specifications
- Désignation:Grade 226 sheets
- Dimensions:8×10"