Drying Chambers, Labconco® | Avantor
Drying Chambers, Labconco®
   Options durables disponibles
Supplier:   Labconco
Drying Chambers, Labconco®
Supplier:  Labconco
   Options durables disponibles
Certificates
Drying Chambers, Labconco®
8 Options Available Below
Product Details & Documents
Designed for use with FreeZone® Freeze Dryers. Provide strength, durability and corrosion resistance.
  • Chambers are available in regular, heated, and clear models
The 12- and 16-port drying chambers are made of Type 304 stainless steel with a clear acrylic top.
Supplied with neoprene valves which accommodate both 1.3 cm (1/2") and 1.9 cm (3/4") adapters. Lids have a neoprene gasket around the edge to assure a leakproof vacuum seal. Models with PTFE-coated interior provide corrosion resistance. The 12.0" diameter clear acrylic chambers (75841-010 and 75841-008) fit all FreeZone® Freeze Dryers and provide visibility into chamber during bulk freeze drying. Accommodate Product Shelves (5 shelves in Tall, 3 in Short). Includes clear acrylic lid with gasket, vacuum release valve and pass-through for cords. Chambers will fit 10.12" or 14.0" diameter condensers. 8" diameter opening to collector on bottom of chamber.
Clear drying chambers (72460-056 and 82010-524) include a clear acrylic lid, gaskets, and electrical pass-through for cords. Provide visibility into chamber during bulk drying. For use with or without product heaters.
Clear Drying Chambers (75841-014 and 75841-012) feature eight valves on top of the chamber, providing a convenient means to attach samples in flasks to a freeze dryer while allowing visibility into the chamber during freeze drying. 12.0" diameter clear acrylic chamber fits all FreeZone® Freeze Dryers and provides visibility into chamber during bulk freeze drying.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 30181-309
  • 30181-311
  • 30181-401
  • 30181-403
  • 82010-512
  • 82010-514
  • 75841-010
  • 75841-008
  • 26672-140
  • 26672-152
  • 75841-034
  • 75841-036
  • 75841-012
  • 75841-014
  • Environmentally Preferable:
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging|Sustainable Materials
  • Sustainable Packaging|Sustainable Materials
  • Sustainable Packaging|Sustainable Materials
  • Sustainable Packaging|Sustainable Materials
  • Sustainable Packaging
  • Sustainable Packaging
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Ø
    Hauteur
    Availability
    Price
  • Clear Chamber, Short
    13.6"
    16.8"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,200.00
    $4,200.00
    Each
       $4,200.00
    La quantité minimale de commande est de 1
    Supplier Number: 7318702
    Spécifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    75841-010
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Clear Chamber, Tall
    13.6"
    24.6"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,290.00
    $5,290.00
    Each
       $5,290.00
    La quantité minimale de commande est de 1
    Supplier Number: 7318700
    Spécifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    75841-008
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • 12-Port Chamber
    8.0"
    9.75"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,220.00
    $4,220.00
    Each
       $4,220.00
    La quantité minimale de commande est de 1
    Supplier Number: 7522800
    Spécifications:
    Product available on GSA Advantage®
      Sustainable Packaging|Sustainable Materials
    More
    Specifications
    Cat. No.
    26672-140
    Environmentally Preferable
    Sustainable Packaging|Sustainable Materials
    Product available on GSA Advantage®
      Sustainable Packaging|Sustainable Materials
  • 16-Port Chamber
    13.0"
    13.0"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,750.00
    $5,750.00
    Each
       $5,750.00
    La quantité minimale de commande est de 1
    Supplier Number: 7522900
    Spécifications:
    Product available on GSA Advantage®
      Sustainable Packaging|Sustainable Materials
    More
    Specifications
    Cat. No.
    26672-152
    Environmentally Preferable
    Sustainable Packaging|Sustainable Materials
    Product available on GSA Advantage®
      Sustainable Packaging|Sustainable Materials
  • 12-Port Chamber, PTFE-Coated
    8.0"
    9.75"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,210.00
    $5,210.00
    Each
       $5,210.00
    La quantité minimale de commande est de 1
    Supplier Number: 7522801
    Spécifications:
      Sustainable Packaging|Sustainable Materials
    More
    Specifications
    Cat. No.
    75841-034
    Environmentally Preferable
    Sustainable Packaging|Sustainable Materials
      Sustainable Packaging|Sustainable Materials
  • 16-Port Chamber, PTFE-Coated
    13.0"
    13.0"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,730.00
    $6,730.00
    Each
       $6,730.00
    La quantité minimale de commande est de 1
    Supplier Number: 7522901
    Spécifications:
      Sustainable Packaging|Sustainable Materials
    More
    Specifications
    Cat. No.
    75841-036
    Environmentally Preferable
    Sustainable Packaging|Sustainable Materials
      Sustainable Packaging|Sustainable Materials
  • Clear Chamber with 8 Valves, Tall
    17.4"
    23.0"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,440.00
    $7,440.00
    Each
       $7,440.00
    La quantité minimale de commande est de 1
    Supplier Number: 7318800
    Spécifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    75841-012
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Clear Chamber with 8 Valves, Short
    17.4"
    15.3"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,400.00
    $6,400.00
    Each
       $6,400.00
    La quantité minimale de commande est de 1
    Supplier Number: 7318802
    Spécifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    75841-014
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging