Manual Pellet Press, Carver
29999-808
: Carver
- Description:Manual Pellet Press
- W×D×H:30.5×35.6×63.5 cm (12×14×25")
- Ram stroke:13 cm (5.1")
- Daylight opening:0 - 14 cm (0 - 5.5")
- Weight:54.4 kg. (120 lbs.)
- Cat. no.:29999-808
- Supplier No.:4350
This compact 12-ton pellet press with 10.2cm (4") diameter platen is perfectly suited for KBr pellet pressing
It can also be used for glove box applications, compound and formula testing, compression strength tests, fluid extractions, and on-site field testing.
: Press includes integral safety shield, buffer plate, 13mm evacuable die, and counter-bored platen for positive die alignment.